5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] 98% mix

Supplier: Ambeed
Warning

A1148024-250MG A1148024-10MG A1148024-100MG A1148024-1G A1148024-50MG A1148024-5MG
76673-738EA 232 USD
76673-738 77595-842 77620-028 77620-030 77620-032 77620-034
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] 98% mix
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)]

Precaución: for Research Use Only

Formula: C₂₉H₁₉NO₁₁
MDL Number: MFCD00037466
CAS Number: 150347-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR