4-Amino-2-(1-methyl-2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 98%

Supplier: Ambeed
Warning

A1147998-100MG A1147998-250MG A1147998-1G
76673-734EA 79 USD
76673-734 76673-736 77570-850
4-Amino-2-(1-methyl-2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 98%
4-Amino-2-(1-methyl-2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₄H₁₃N₃O₄
MDL Number: MFCD31703382
CAS Number: 1352827-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR