4,4'-(((Ethane-1,2-diylbis(oxy))bis(ethane-2,1-diyl))bis(azanediyl))bis(2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione) 98%

Supplier: Ambeed
Warning

Synonyms: CRBN PROTAC compound I

A1147988-10MG A1147988-1MG A1147988-25MG A1147988-50MG A1147988-5MG
76673-724EA 180.36 USD
76673-724 76673-726 76673-728 76673-730 76673-732
4,4'-(((Ethane-1,2-diylbis(oxy))bis(ethane-2,1-diyl))bis(azanediyl))bis(2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione) 98%
4,4'-(((Ethane-1,2-diylbis(oxy))bis(ethane-2,1-diyl))bis(azanediyl))bis(2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione)

Precaución: for Research Use Only

Formula: C₃₂H₃₂N₆O₁₀
MW: 660.6307 g/mol
MDL Number: MFCD31813818
CAS Number: 2244520-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR