Saccharin sodium hydrate 97%

Supplier: Ambeed
Warning

A1147966-100G A1147966-25G A1147966-500G
76673-716EA 31 USD
76673-716 76673-718 76673-720
Saccharin sodium hydrate 97%
Saccharin sodium hydrate

Precaución: for Research Use Only

Formula: C₇H₅NNaO₃S·xH₂O
MDL Number: MFCD00149605
CAS Number: 82385-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR