(S)-2-(Piperazin-2-yl)acetonitrile dihydrochloride 97%

Supplier: Ambeed
Warning

A1147954-100MG A1147954-1G A1147954-250MG A1147954-25G A1147954-5G
76673-706EA 16.98 USD
76673-706 76673-708 76673-710 76673-712 76673-714
(S)-2-(Piperazin-2-yl)acetonitrile dihydrochloride 97%
(S)-2-(Piperazin-2-yl)acetonitrile dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃Cl₂N₃
MW: 198.0935 g/mol
MDL Number: MFCD32173174
CAS Number: 2158301-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR