(3aS,3a'S,8aR,8a'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole) ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (3aS,3a'S,8aR,8a'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole)(97%)

A1147899-100MG A1147899-1G A1147899-250MG
76673-674EA 90.18 USD
76673-674 76673-676 76673-678
(3aS,3a'S,8aR,8a'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole) ≥98%, ee 99%
(3aS,3a'S,8aR,8a'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole)

Precaución: for Research Use Only

Formula: C₄₃H₄₆N₂O₂
MW: 622.8376 g/mol
CAS Number: 1435467-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR