(S,S)-4,4'-Diisopropyl-4,5,4',5'-tetrahydro[2.2]bioxazolyl ≥98%, ee 99%

Supplier: Ambeed
Warning

A1147895-100MG A1147895-1G A1147895-250MG A1147895-5G
76673-664EA 19 USD
76673-664 76673-666 76673-668 77620-026
(S,S)-4,4'-Diisopropyl-4,5,4',5'-tetrahydro[2.2]bioxazolyl ≥98%, ee 99%
(S,S)-4,4'-Diisopropyl-4,5,4',5'-tetrahydro[2.2]bioxazolyl

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₂
MW: 224.2994 g/mol
MDL Number: MFCD32633535
CAS Number: 131833-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR