(S)-3,3'-Bis[4-(trifluoromethyl)phenyl]-[1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%

Supplier: Ambeed
Warning

A1147888-100MG A1147888-1G A1147888-250MG A1147888-5G
76673-638EA 63.66 USD
76673-638 76673-640 76673-642 76673-644
(S)-3,3'-Bis[4-(trifluoromethyl)phenyl]-[1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%
(S)-3,3'-Bis[4-(trifluoromethyl)phenyl]-[1,1'-binaphthalene]-2,2'-diol

Precaución: for Research Use Only

CAS Number: 1621066-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR