(11bS)-4-Hydroxy-2,6-bis[2,3,4,5,6-pentafluorophenyl]-4-oxide-dinaphtho[2,1-d:1,2-f][1,3,2]dioxaphosphepin 98%

Supplier: Ambeed
Warning

A1147884-100MG A1147884-1G A1147884-250MG
76673-616EA 78.51 USD
76673-616 76673-618 76673-620
(11bS)-4-Hydroxy-2,6-bis[2,3,4,5,6-pentafluorophenyl]-4-oxide-dinaphtho[2,1-d:1,2-f][1,3,2]dioxaphosphepin 98%
(11bS)-4-Hydroxy-2,6-bis[2,3,4,5,6-pentafluorophenyl]-4-oxide-dinaphtho[2,1-d:1,2-f][1,3,2]dioxaphosphepin

Precaución: for Research Use Only

MDL Number: MFCD31720587
CAS Number: 1882075-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR