1-Methyl-9H-carbazole 98%

Supplier: Ambeed
Warning

A1147718-100MG A1147718-1G A1147718-250MG
76673-566EA 13 USD
76673-566 76673-568 76673-570
1-Methyl-9H-carbazole 98%
1-Methyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₃H₁₁N
MDL Number: MFCD18450170
CAS Number: 6510-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR