2-Phenyl-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 98%

Supplier: Ambeed
Danger

A1147623-1G A1147623-250MG A1147623-25G A1147623-5G
76673-550EA 41 USD
76673-550 76673-552 76673-554 76673-556
2-Phenyl-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 98%
2-Phenyl-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Precaución: for Research Use Only

Formula: C₁₁H₁₂BNO₄
MDL Number: MFCD11215250
CAS Number: 109674-45-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR