N-Carbamimidoyl-2-methyl-4,5-bis(methylsulfonyl)benzamide 99%

Supplier: Ambeed
Warning

A1147312-1MG A1147312-5MG
76673-522EA 58 USD
76673-522 76673-524
N-Carbamimidoyl-2-methyl-4,5-bis(methylsulfonyl)benzamide 99%
N-Carbamimidoyl-2-methyl-4,5-bis(methylsulfonyl)benzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₃O₅S₂
MDL Number: MFCD30533663
CAS Number: 187870-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR