Magnesium sulfate monohydrate 99.5%

Supplier: Ambeed
Warning

A1147147-25G A1147147-100G
76673-508EA 5 USD
76673-508 77620-010
Magnesium sulfate monohydrate 99.5%
Magnesium sulphate monohydrate

Precaución: for Research Use Only

Formula: MgSO₄·1H₂O
MDL Number: MFCD00149787
CAS Number: 14168-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR