EDTA disodium zinc salt tetrahydrate 98% for complexometry

Supplier: Ambeed
Warning

Synonyms: Disodium zinc ethylenediaminetetraacetate tetrahydrate, Ethylenediaminetetraacetic Acid Disodium Zinc Salt tetraHydrate, EDTA-Na₂Zn

A1147126-1000G A1147126-100G A1147126-500G
76673-502EA 53 USD
76673-502 76673-504 76673-506
EDTA disodium zinc salt tetrahydrate 98% for complexometry
EDTA disodium zinc salt tetrahydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂Na₂O₈Zn·4H₂O
MDL Number: MFCD00151728
CAS Number: 176736-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR