Methyl 4-bromo-5-chloropicolinate 97%

Supplier: Ambeed
Warning

A1147087-100MG A1147087-1G A1147087-250MG
76673-494EA 93.36 USD
76673-494 76673-496 76673-498
Methyl 4-bromo-5-chloropicolinate 97%
Methyl 4-bromo-5-chloropicolinate

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MW: 250.4771 g/mol
MDL Number: MFCD18257373
CAS Number: 1256834-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR