(4-(Aminomethyl)pyridin-2-yl)methanol hydrochloride 97%

Supplier: Ambeed
Warning

A1146946-100MG A1146946-1G A1146946-5G
76673-478EA 57.29 USD
76673-478 76673-480 76673-482
(4-(Aminomethyl)pyridin-2-yl)methanol hydrochloride 97%
(4-(Aminomethyl)pyridin-2-yl)methanol hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂O
MW: 174.6300 g/mol
CAS Number: 1844884-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR