N,N,N',N'-Tetra([1,1'-biphenyl]-4-yl)[1,1':4',1''-terphenyl]-4,4''-diamine 98%

Supplier: Ambeed
Warning

A1146924-100MG A1146924-1G A1146924-250MG A1146924-5G
76673-472EA 18 USD
76673-472 76673-474 76673-476 77865-903
N,N,N',N'-Tetra([1,1'-biphenyl]-4-yl)[1,1':4',1''-terphenyl]-4,4''-diamine 98%
N,N,N',N'-Tetra([1,1'-biphenyl]-4-yl)[1,1':4',1''-terphenyl]-4,4''-diamine

Precaución: for Research Use Only

Formula: C₆₆H₄₈N₂
CAS Number: 952431-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR