4,7-Bis(5-n-octyl-2-thienyl)-2,1,3-benzothiadiazole 98%

Supplier: Ambeed
Warning

Synonyms: 4,7-Bis(5-octyl-2-thienyl)-2,1,3-benzothiadiazole

A1146920-100MG A1146920-1G A1146920-250MG A1146920-50MG
76673-464EA 62.6 USD
76673-464 76673-466 76673-468 76673-470
4,7-Bis(5-n-octyl-2-thienyl)-2,1,3-benzothiadiazole 98%
4,7-Bis(5-n-octyl-2-thienyl)-2,1,3-benzothiadiazole

Precaución: for Research Use Only

Formula: C₃₀H₄₀N₂S₃
MW: 524.84 g/mol
MDL Number: MFCD28976158
CAS Number: 1171974-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR