Manganesebis(trifluoromethylsulfonyl)imide 97%

Supplier: Ambeed
Danger

Synonyms: Manganese(II) triflimide, Manganese(II) Bis(trifluoromethanesulphonyl)imide, Manganese bis(trifluoromethanesulphonyl)imide, Bis(trifluoromethanesulphonyl)imide manganese(II) salt

A1146864-1G A1146864-250MG A1146864-5G A1146864-25G
76673-458EA 50.93 USD
76673-458 76673-460 76673-462 77595-818
Manganesebis(trifluoromethylsulfonyl)imide 97%
Manganesebis(trifluoromethylsulfonyl)imide

Precaución: for Research Use Only

Formula: C₄F₁₂MnN₂O₈S₄
MW: 615.21 g/mol
MDL Number: MFCD23380173
CAS Number: 207861-55-0
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR