(2R,4R)-2-(2-Chlorophenyl)-3-(4-(3,5-dimethylisoxazol-4-yl)benzoyl)thiazolidine-4-carboxylic acid 99+%
Supplier: Ambeed
|
Warning
|
Synonyms:
TUG-1375
A1146800-10MG
A1146800-1MG
A1146800-25MG
A1146800-5MG
76673-450EA
168.69
USD
76673-450
76673-452
76673-454
76673-456
(2R,4R)-2-(2-Chlorophenyl)-3-(4-(3,5-dimethylisoxazol-4-yl)benzoyl)thiazolidine-4-carboxylic acid 99+%
(2R,4R)-2-(2-Chlorophenyl)-3-(4-(3,5-dimethylisoxazol-4-yl)benzoyl)thiazolidine-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₂₂H₁₉ClN₂O₄S MW: 442.9153 g/mol |
MDL Number:
MFCD32062757 CAS Number: 2247372-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...