(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I) 98%

Supplier: Ambeed
Warning

A1146758-100MG A1146758-1G A1146758-250MG A1146758-5G
76673-442EA 25 USD
76673-442 76673-444 76673-446 76673-448
(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I) 98%
(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I)

Precaución: for Research Use Only

Formula: C₃₁H₂₃CuF₃N₂P
MW: 575.05 g/mol
MDL Number: MFCD28386111
CAS Number: 1334890-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR