Dimethyl 1,1'-ferrocenedicarboxylate 98%

Supplier: Ambeed
Warning

A1146568-1G A1146568-250MG A1146568-25G A1146568-5G
76673-434EA 44 USD
76673-434 76673-436 76673-438 76673-440
Dimethyl 1,1'-ferrocenedicarboxylate 98%
Dimethyl 1,1'-ferrocenedicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₆FeO₄
MDL Number: MFCD00009950
CAS Number: 1273-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR