Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]Cobalt(II) 98%
Supplier: Ambeed
|
Warning
|
A1146440-1G
A1146440-250MG
76673-430EA
85.94
USD
76673-430
76673-432
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]Cobalt(II) 98%
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]Cobalt(II)
Precaución: for Research Use Only
Formula:
C₃₉H₃₂Cl₂CoOP₂ MW: 708.4575 g/mol |
MDL Number:
MFCD31718123 CAS Number: 898807-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...