Tris(4-bromophenyl)phosphane 95%

Supplier: Ambeed
Warning

A1146438-100MG A1146438-1G A1146438-250MG A1146438-5G
76673-422EA 18.04 USD
76673-422 76673-424 76673-426 76673-428
Tris(4-bromophenyl)phosphane 95%
Tris(4-bromophenyl)phosphane

Precaución: for Research Use Only

Formula: C₁₈H₁₂Br₃P
MW: 498.9736 g/mol
MDL Number: MFCD02690473
CAS Number: 29949-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR