1,4-Bis(diphenylphosphanyl)benzene 98%

Supplier: Ambeed
Warning

A1146428-1G A1146428-250MG A1146428-5G
76673-418EA 108.22 USD
76673-418 76673-420 77566-024
1,4-Bis(diphenylphosphanyl)benzene 98%
1,4-Bis(diphenylphosphanyl)benzene

Precaución: For research use only.

Formula: C₃₀H₂₄P₂
MW: 446.4591 g/mol
MDL Number: MFCD00014082
CAS Number: 1179-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR