Methyl 1-bromo-2-naphthoate 98%

Supplier: Ambeed
Warning

A1146424-250MG A1146424-1G A1146424-5G
76673-416EA 25.47 USD
76673-416 77595-814 77619-996
Methyl 1-bromo-2-naphthoate 98%
Methyl 1-bromo-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₂H₉BrO₂
MW: 265.1000 g/mol
MDL Number: MFCD06203695
CAS Number: 89555-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR