2-Bromo-11H-benzo[b]fluoren-11-one 98%

Supplier: Ambeed
Warning

A1145842-1G A1145842-5G
76673-406EA 20 USD
76673-406 76673-408
2-Bromo-11H-benzo[b]fluoren-11-one 98%
2-Bromo-11H-benzo[b]fluoren-11-one

Precaución: for Research Use Only

Formula: C₁₇H₉BrO
MDL Number: MFCD32012308
CAS Number: 1923756-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR