DIM-C-pPhCO2Me 99+%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-(di(1H-indol-3-yl)methyl)benzoate

A1145756-10MG A1145756-1MG A1145756-25MG A1145756-50MG A1145756-5MG
76673-396EA 33.95 USD
76673-396 76673-398 76673-400 76673-402 76673-404
DIM-C-pPhCO2Me 99+%
DIM-C-pPhCO2Me

Precaución: for Research Use Only

Formula: C₂₅H₂₀N₂O₂
MW: 380.45 g/mol
MDL Number: MFCD31697720
CAS Number: 151358-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR