3-Ethyl-6-(4-hydroxypiperidine-1-carbonyl)-5-methyl-1-(3,3,3-trifluoropropyl)thieno[2,3-d]pyrimidine-2,4(1H,3H)-dione 98%

Supplier: Ambeed
Warning

Synonyms: BAY-545

A1145746-1MG A1145746-5MG
76673-388EA 43.5 USD
76673-388 76673-390
3-Ethyl-6-(4-hydroxypiperidine-1-carbonyl)-5-methyl-1-(3,3,3-trifluoropropyl)thieno[2,3-d]pyrimidine-2,4(1H,3H)-dione 98%
3-Ethyl-6-(4-hydroxypiperidine-1-carbonyl)-5-methyl-1-(3,3,3-trifluoropropyl)thieno[2,3-d]pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₈H₂₂F₃N₃O₄S
MW: 433.45 g/mol
CAS Number: 1699717-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR