(S)-6-(4-(5-(3,5-Difluorophenyl)-4,5-dihydro-1H-pyrazole-1-carbonyl)piperidin-1-yl)pyrimidine-4-carbonitrile 99+%

Supplier: Ambeed
Warning

Synonyms: GSK547

A1145742-10MG A1145742-1MG A1145742-25MG A1145742-5MG A1145742-100MG A1145742-50MG A1145742-250MG
76673-380EA 140 USD
76673-380 76673-382 76673-384 76673-386 77619-992 77619-994 77865-891
(S)-6-(4-(5-(3,5-Difluorophenyl)-4,5-dihydro-1H-pyrazole-1-carbonyl)piperidin-1-yl)pyrimidine-4-carbonitrile 99+%
(S)-6-(4-(5-(3,5-Difluorophenyl)-4,5-dihydro-1H-pyrazole-1-carbonyl)piperidin-1-yl)pyrimidine-4-carbonitrile

Precaución: for Research Use Only

Formula: C₂₀H₁₈F₂N₆O
MDL Number: MFCD32062767
CAS Number: 2226735-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR