2-(5-Phenyl-4-(pyridin-4-yl)-1H-1,2,3-triazol-1-yl)-N-(5-phenylpyridin-2-yl)acetamide 99+%

Supplier: Ambeed
Warning

A1145740-10MG A1145740-1MG A1145740-25MG A1145740-5MG
76673-372EA 50 USD
76673-372 76673-374 76673-376 76673-378
2-(5-Phenyl-4-(pyridin-4-yl)-1H-1,2,3-triazol-1-yl)-N-(5-phenylpyridin-2-yl)acetamide 99+%
2-(5-Phenyl-4-(pyridin-4-yl)-1H-1,2,3-triazol-1-yl)-N-(5-phenylpyridin-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₂₆H₂₀N₆O
MDL Number: MFCD31697718
CAS Number: 2074607-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR