3-(1-Aminoethyl)pyridin-2(1H)-one hydrochloride 97%

Supplier: Ambeed
Warning

A1145688-100MG A1145688-250MG A1145688-5G A1145688-1G
76673-358EA 24 USD
76673-358 76673-360 76673-362 77865-887
3-(1-Aminoethyl)pyridin-2(1H)-one hydrochloride 97%
3-(1-Aminoethyl)pyridin-2(1H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂O
MW: 174.6280 g/mol
CAS Number: 2411219-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR