N'-((3-Hydroxy-5-(hydroxymethyl)-2-methylpyridin-4-yl)methylene)isonicotinohydrazide 98%

Supplier: Ambeed
Warning

Synonyms: Pyridoxal isonicotinoyl hydrazone

A1145678-10MG A1145678-1MG A1145678-50MG
76673-352EA 32 USD
76673-352 76673-354 76673-356
N'-((3-Hydroxy-5-(hydroxymethyl)-2-methylpyridin-4-yl)methylene)isonicotinohydrazide 98%
N'-((3-Hydroxy-5-(hydroxymethyl)-2-methylpyridin-4-yl)methylene)isonicotinohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₄O₃
MW: 286.2860 g/mol
MDL Number: MFCD00763290
CAS Number: 737-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR