2-((1-Benzyl-1H-pyrazol-4-yl)oxy)pyrido[3,4-d]pyrimidin-4(3H)-one 98%

Supplier: Ambeed
Warning

Synonyms: GSK467

A1145674-10MG A1145674-1MG A1145674-25MG A1145674-5MG
76673-344EA 525.15 USD
76673-344 76673-346 76673-348 76673-350
2-((1-Benzyl-1H-pyrazol-4-yl)oxy)pyrido[3,4-d]pyrimidin-4(3H)-one 98%
2-((1-Benzyl-1H-pyrazol-4-yl)oxy)pyrido[3,4-d]pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₁₇H₁₃N₅O₂
MW: 319.32 g/mol
MDL Number: MFCD32067913
CAS Number: 1628332-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR