3-Methoxy-N-(1-methylpiperidin-4-yl)-4-((4-((3-oxo-2,3-dihydro-1H-inden-4-yl)oxy)-5-(trifluoromethyl)pyrimidin-2-yl)amino)benzamide 98%

Supplier: Ambeed
Warning

A1145670-10MG A1145670-1MG A1145670-25MG A1145670-5MG
76673-336EA 83 USD
76673-336 76673-338 76673-340 76673-342
3-Methoxy-N-(1-methylpiperidin-4-yl)-4-((4-((3-oxo-2,3-dihydro-1H-inden-4-yl)oxy)-5-(trifluoromethyl)pyrimidin-2-yl)amino)benzamide 98%
3-Methoxy-N-(1-methylpiperidin-4-yl)-4-((4-((3-oxo-2,3-dihydro-1H-inden-4-yl)oxy)-5-(trifluoromethyl)pyrimidin-2-yl)amino)benzamide

Precaución: for Research Use Only

Formula: C₂₈H₂₈F₃N₅O₄
MDL Number: MFCD32062817
CAS Number: 1227948-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR