Acrizanib 98%

Supplier: Ambeed
Warning

Synonyms: N-(1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl)-5-((6-((methylamino)methyl)pyrimidin-4-yl)oxy)-1H-indole-1-carboxamide

A1145660-1MG A1145660-5MG
76673-314EA 73.21 USD
76673-314 76673-316
Acrizanib 98%
Acrizanib

Precaución: for Research Use Only

Formula: C₂₀H₁₈F₃N₇O₂
MW: 445.4 g/mol
MDL Number: MFCD31715417
CAS Number: 1229453-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR