4-((2-(6-(4-Methylpiperazine-1-carbonyl)naphthalen-2-yl)ethyl)amino)quinazoline-6-carbonitrile 98%

Supplier: Ambeed
Warning

A1145598-1MG A1145598-5MG
76673-292EA 21 USD
76673-292 76673-294
4-((2-(6-(4-Methylpiperazine-1-carbonyl)naphthalen-2-yl)ethyl)amino)quinazoline-6-carbonitrile 98%
4-((2-(6-(4-Methylpiperazine-1-carbonyl)naphthalen-2-yl)ethyl)amino)quinazoline-6-carbonitrile

Precaución: for Research Use Only

Formula: C₂₇H₂₆N₆O
MDL Number: MFCD31544331
CAS Number: 1449228-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR