5-Bromo-5'-phenyl-3'H-spiro[indoline-3,2'-[1,3,4]thiadiazol]-2-one 98%

Supplier: Ambeed
Warning

A1145370-10MG A1145370-1MG A1145370-25MG A1145370-50MG A1145370-5MG
76673-280EA 233.4 USD
76673-280 76673-282 76673-284 76673-286 76673-288
5-Bromo-5'-phenyl-3'H-spiro[indoline-3,2'-[1,3,4]thiadiazol]-2-one 98%
5-Bromo-5'-phenyl-3'H-spiro[indoline-3,2'-[1,3,4]thiadiazol]-2-one

Precaución: for Research Use Only

Formula: C₁₅H₁₀BrN₃OS
MW: 360.2284 g/mol
MDL Number: MFCD00770315
CAS Number: 297180-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR