5-Phenyl-5,8-dihydroindolo[2,3-c]carbazole 98%

Supplier: Ambeed
Warning

A1145242-100MG A1145242-1G A1145242-250MG
76673-268EA 38 USD
76673-268 76673-270 76673-272
5-Phenyl-5,8-dihydroindolo[2,3-c]carbazole 98%
5-Phenyl-5,8-dihydroindolo[2,3-c]carbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₆N₂
CAS Number: 1637752-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR