(R)-12-((S)-7,8-Difluoro-6,11-dihydrodibenzo[b,e]thiepin-11-yl)-7-hydroxy-3,4,12,12a-tetrahydro-1H-[1,4]oxazino[3,4-c]pyrido[2,1-f][1,2,4]triazine-6,8-dione 97%

Supplier: Ambeed
Warning

Synonyms: Baloxavir

A1145224-1G A1145224-250MG A1145224-5G A1145224-100MG
76673-262EA 283.27 USD
76673-262 76673-264 76673-266 77595-790
(R)-12-((S)-7,8-Difluoro-6,11-dihydrodibenzo[b,e]thiepin-11-yl)-7-hydroxy-3,4,12,12a-tetrahydro-1H-[1,4]oxazino[3,4-c]pyrido[2,1-f][1,2,4]triazine-6,8-dione 97%
(R)-12-((S)-7,8-Difluoro-6,11-dihydrodibenzo[b,e]thiepin-11-yl)-7-hydroxy-3,4,12,12a-tetrahydro-1H-[1,4]oxazino[3,4-c]pyrido[2,1-f][1,2,4]triazine-6,8-dione

Precaución: for Research Use Only

Formula: C₂₄H₁₉F₂N₃O₄S
MW: 483.49 g/mol
MDL Number: MFCD31619273
CAS Number: 1985605-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR