1-Isopropylpyrazole-4-boronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1-Isopropyl-1H-pyrazol-4-yl)boronic acid
A1145118-100MG
A1145118-250MG
A1145118-1G
76673-254EA
35.01
USD
76673-254
77549-234
77570-828
1-Isopropylpyrazole-4-boronic acid 97%
1-Isopropylpyrazole-4-boronic acid
Precaución: for Research Use Only
Formula:
C₆H₁₁BN₂O₂ MW: 153.975 g/mol |
MDL Number:
MFCD16036118 CAS Number: 1201643-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...