1H-Indazole-5-boronic acid 98%

Supplier: Ambeed
Warning

A1145100-100MG A1145100-1G A1145100-250MG A1145100-5G A1145100-25G
76673-246EA 16.98 USD
76673-246 76673-248 76673-250 76673-252 77595-788
1H-Indazole-5-boronic acid 98%
1H-Indazole-5-boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BN₂O₂
MW: 161.9537 g/mol
MDL Number: MFCD06739151
CAS Number: 338454-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR