Methyl 1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanecarboxylate 97%
Supplier: Ambeed
|
Warning
|
A1145088-100MG
A1145088-1G
76673-228EA
50.93
USD
76673-228
76673-230
Methyl 1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanecarboxylate 97%
Methyl 1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanecarboxylate
Precaución: for Research Use Only
Formula:
C₁₇H₂₃BO₄ MW: 302.1731 g/mol |
MDL Number:
MFCD18383383 CAS Number: 1396007-85-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...