5-Vinylisophthalic acid 97% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A1145082-250MG A1145082-5G
76673-224EA 28 USD
76673-224 76673-226
5-Vinylisophthalic acid 97% stabilized
5-Vinylisophthalic acid

Precaución: for Research Use Only

Formula: C₁₀H₈O₄
CAS Number: 1041374-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR