(4-Chloro-2-(methoxycarbonyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-(methoxycarbonyl)phenylboronic acid

A1145060-1G A1145060-250MG A1145060-5G
76673-214EA 24 USD
76673-214 76673-216 77865-869
(4-Chloro-2-(methoxycarbonyl)phenyl)boronic acid 98%
(4-Chloro-2-(methoxycarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BClO₄
MW: 214.4100 g/mol
MDL Number: MFCD25459257
CAS Number: 1612256-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR