5-Chloro-2-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)benzoic acid methyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

A1145058-100MG A1145058-1G A1145058-250MG A1145058-5G
76673-206EA 18.04 USD
76673-206 76673-208 76673-210 76673-212
5-Chloro-2-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)benzoic acid methyl ester 95%
5-Chloro-2-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BClO₄
MW: 296.5543 g/mol
MDL Number: MFCD16996244
CAS Number: 866625-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR