7-Azaindole-5-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (1H-Pyrrolo[2,3-b]pyridin-5-yl)boronic acid, {1H-pyrrolo[2,3-b]pyridin-5-yl}boronic acid

A1145036-100MG A1145036-1G A1145036-250MG
76673-192EA 42.44 USD
76673-192 76673-194 77592-222
7-Azaindole-5-boronic acid 97%
7-Azaindole-5-boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BN₂O₂
MW: 161.96 g/mol
MDL Number: MFCD08234881
CAS Number: 944059-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR