Dicyclohexyl(3-isopropoxy-2',4',6'-triisopropyl-[1,1'-biphenyl]-2-yl)phosphine 97%

Supplier: Ambeed
Warning

A1144952-1G A1144952-250MG A1144952-100MG
76673-186EA 891.16 USD
76673-186 77566-016 77619-962
Dicyclohexyl(3-isopropoxy-2',4',6'-triisopropyl-[1,1'-biphenyl]-2-yl)phosphine 97%
Dicyclohexyl(3-isopropoxy-2',4',6'-triisopropyl-[1,1'-biphenyl]-2-yl)phosphine

Precaución: For research use only.

Formula: C₃₆H₅₅OP
MW: 534.795 g/mol
MDL Number: MFCD31619192
CAS Number: 2118959-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR