Retro-2 cycl 98%

Supplier: Ambeed
Warning

Synonyms: RN 1-001, Retro-2, 2-(5-Methylthiophen-2-yl)-3-phenyl-2,3-dihydroquinazolin-4(1H)-one

A1144862-10MG A1144862-1MG A1144862-5MG
76673-180EA 115.64 USD
76673-180 76673-182 76673-184
Retro-2 cycl 98%
Retro-2 cycl

Precaución: for Research Use Only

Formula: C₁₉H₁₆N₂OS
MW: 320.41 g/mol
MDL Number: MFCD28124399
CAS Number: 1429192-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR