2-Methyl-1,2,3,4-tetrahydroisoquinoline-6,7-diol hydrobromide 97%

Supplier: Ambeed
Warning

A1144310-100MG A1144310-1G A1144310-250MG A1144310-50MG
76673-156EA 85 USD
76673-156 76673-158 76673-160 76673-162
2-Methyl-1,2,3,4-tetrahydroisoquinoline-6,7-diol hydrobromide 97%
2-Methyl-1,2,3,4-tetrahydroisoquinoline-6,7-diol hydrobromide

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrNO₂
CAS Number: 57553-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR